Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ LDPE Carboy
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
10 L
20 L
50 L
Description
- Store and transport reagents with 10 L FISHERBRAND LDPE, Carboy, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands.
- These carboys accept 13.5 rubber stoppers for an alternative sealing method.
- Molded-in graduations of 5 L and 1 gal. increments
- Leakproof
- Includes:White polypropylene screw cap and 83B closure
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 20 L |
| Color | Natural |
| Disposable | Reusable |
| Material | LDPE |
| Mouth Style | Wide |
| Product Type | Carboy |
| Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction