Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mettler Toledo™ Mettler Toledo™ Micro pH Electrode: LE422
Shop All METTLER TOLEDO Products
Click to view available options
Connector Types:
BNC
Description
- Ag/AgCl reference system
- Gel electrolyte filling
- Low maintenance
- U-membrane glass
- Glass shaft
- Ceramic frit
- LE cap at electrode head
- pH Range: 0 to 14
- Zero Point (pH): 7.00±0.25
- Temperature Range: 0° to 80°C
- Membrane Resistance: <250MOhm
- Lower Shaft Length: 90mm
- Lower Shaft Dia.: 4.3mm
- Cable Length: 1m
- Connector: BNC
Recommended Storage
Store for short term in 3mol/L KCl in watering cap and for long-term storage, store in 3mol/L KCl, buffer pH 4 or 7.
Specifications
Specifications
| Description | pH electrode LE422 |
| Connector Types | BNC |
| Material | Glass |
| pH Range | 0 to 14 |
| Filling Solution Type | Gel |
| For Use With (Application) | Measuring in small or narrow neck vessels or test tubes. |
| Internal Reference | Ag/AgCl |
| Model | pH electrode LE422 |
| Storage Requirements | Store for short term in 3mol/L KCl in watering cap and for long-term storage store in 3mol/L KCl, buffer pH 4 or 7. |
| Temperature (Metric) Range | 0°C to 80°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction