Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate, 10% (w/v), Ricca Chemical
SDP

Catalog No. 4293432 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7732-18-5, 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Physical Form Liquid
Quantity 1 L
Percent Purity 10% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Lead Acetate
ShelfLife 6 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.