Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach™ Lead Acid Battery, 12 V

Catalog No. 029380009
Click to view available options
Style:
3 Pin Connector
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with SD900/AS950 Samplers, compatible with charger 8753500

TRUSTED_SUSTAINABILITY

Specifications

Product Type Lead Acid Battery
Style 3 Pin Connector
For Use With (Application) SD900/AS950 Samplers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.