Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Carbonate, Powder, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18603763 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

L1045, 598-63-0, PbCO3

Spectrum™ Chemical Lead Carbonate, Powder, Reagent, ACS is a chemical compound that is created industrially from lead acetate and carbon dioxide. It is an important pigment used in paints, putty, and ceramics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
IUPAC Name λ²-lead(2+) carbonate
SMILES [Pb++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 6 x 500 g
Packaging Poly Bottle
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.