Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Dioxide, Powder, 97%, Spectrum™ Chemical
SDP

Catalog No. 18611232 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-232 50 kg Fiber Drum
18-611-231 25 g Amber Glass Bottle
18-611-228 100 g Poly Bottle
18-611-233 500 g Poly Bottle
18-611-230 2.5 kg Poly Pail
18-611-229 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-232 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L106350KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 1309-60-0

Specifications

CAS Max % 1
Quantity 50 kg
Percent Purity 97%
Grade Reagent
Packaging Fiber Drum
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.