Learn More
Lead ICP Standard, 10,000 ppm Pb in 5% HNO3, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 10,000ppm Pb |
| CAS Min % | 84.0, 12.69, 0.85 |
| CAS Max % | 88.0, 13.21, 0.88 |
| Equivalence | 1 mL = 10 mg Pb |
| Physical Form | Liquid |
| Color | Colorless |
| pH | < 2 |
| Type | Standard |
| Quantity | 100 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.