Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) 2,4-pentanedionate

Catalog No. AA8985809
Click to view available options
Quantity:
10 g
50 g

Catalog No. AA8985809

Supplier: Thermo Scientific Chemicals 08985809

Only null left
Add to Cart

CAS: 15282-88-9 | C10H14O4Pb | 405.418 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15282-88-9
Molecular Formula C10H14O4Pb
Molecular Weight (g/mol) 405.418
MDL Number MFCD00013499
InChI Key UNNUWSQNTAFLDC-SYWGCQIGSA-L
Synonym lead ii acetylacetonate, 2,4-pentanedione lead ii derivative
PubChem CID 53393517
IUPAC Name bis[[(E)-4-oxopent-2-en-2-yl]oxy]lead
SMILES CC(=CC(=O)C)O[Pb]OC(=CC(=O)C)C

Specifications

Melting Point 141°C to 144°C
Odor Odorless
Quantity 10 g
UN Number UN3467
Sensitivity Hygroscopic
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 405.41
Physical Form Crystalline
Chemical Name or Material Lead (II) 2,4-pentanedionate
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
EINECSNumber 239-323-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.