Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) acetate, basic, ACS

Catalog No. p-7050290
Click to view available options
Quantity:
500 g

CAS: 1335-32-6

Lead(II) acetate is used in the production of sweeteners, cosmetics, astringent, used in the cleaning and maintenance of stainless steel firearm suppressors (silencers) and compensators, and also used in making of slow matches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1335-32-6
MDL Number MFCD00013041
Synonym Lead subacetate

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula Pb(OOCCH3)2·2Pb(OH)2
UN Number UN1616
Merck Index 14,5419
Solubility Information Soluble in water.
Formula Weight 807.71
Grade ACS
Sensitivity Air Sensitive
Chemical Name or Material Lead(II) acetate, Basic
Hazard Category H351-H360Df-H373-H500
Hazard Statement GHS H Statement
H360-H351-H373
May damage fertility or the unborn child.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber 215-630-3
RTECSNumber OF8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.