Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) acetate, basic

Catalog No. AAA167630E
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA167630E 2500 g
AAA1676336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA167630E Supplier Thermo Scientific Chemicals Supplier No. A167630E
Add to Cart

CAS: 1335-32-6 | C4H6O4Pb·2Pb(OH)2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1335-32-6
Molecular Formula C4H6O4Pb·2Pb(OH)2
MDL Number MFCD00013041
Synonym Lead subacetate

Specifications

Linear Formula Pb(OOCCH3)2·2Pb(OH)2
Quantity 2500 g
UN Number UN1616
Sensitivity Air sensitive
Merck Index 14,5419
Formula Weight 807.72
Chemical Name or Material Lead(II) acetate, basic
Hazard Category H351-H360Df-H373-H500
Hazard Statement GHS H Statement
H360-H351-H373
May damage fertility or the unborn child.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber 215-630-3
RTECSNumber OF8750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.