Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) carbonate, ACS

Catalog No. AA4011957
Change view
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4011957 4 x 500 g
AA4011922 100 g
AA4011936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4011957 Supplier Thermo Scientific Chemicals Supplier No. 04011957
Only null left
Add to Cart

Maximum level of impurities: Insoluble in dilute acetic acid 0.02%, Cl 0.002%, Nitrate and nitrite (as NO3) P.T. (limit about 0.005%), Ca 0.01%, K 0.02%, Na 0.05%, Cd 0.002%, Fe 0.005%, Zn 0.003% | CAS: 598-63-0 | CO3Pb | 267.20 g/mol

Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
MDL Number MFCD00064631
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
PubChem CID 11727
SMILES [Pb++].[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 4 x 500 g
Solubility Information Soluble in acid and alkali. Insoluble in water,alcohol and ammonia.
IUPAC Name λ²-lead(2+) carbonate
Formula Weight 267.2
Grade ACS
Odor Odorless
Appearance White powder
Chemical Name or Material Lead(II) carbonate, For analysis ACS
Hazard Category H302+H332-H350-H360Df-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation DOT Class: 9; Packaging Group: III
EINECSNumber 209-943-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.