Learn More
Lead(II) carbonate, ACS, Thermo Scientific Chemicals
Description
Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Solubility Information | Soluble in acid and alkali. Insoluble in water,alcohol and ammonia. |
IUPAC Name | lead(2+);carbonate |
Formula Weight | 267.2 |
Grade | ACS |
Odor | Odorless |
Appearance | White powder |
Density | 6.6g/cm3 at 20°C |
Show More |
Safety and Handling
Hazard Category | H302+H332-H350-H360Df-H373 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | DOT Class: 9; Packaging Group: III |
EINECSNumber | 209-943-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.