Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) carbonate, basic, extra pure

Catalog No. AC222710050
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222710050 5 g Glass Bottle
AC222715000 500 g Plastic bottle
AC222710025 2.5 kg Plastic bottle
AC222710100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC222710050 Supplier Thermo Scientific Chemicals Supplier No. 222710050
Add to Cart

CAS: 1319-46-6 | C2H2O8Pb3 | 775.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
Molecular Formula C2H2O8Pb3
Molecular Weight (g/mol) 775.59
MDL Number MFCD00078155

Specifications

Melting Point 400.0°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula (PbCO3)2·Pb(OH)2
Merck Index 15, 1002
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 775.59
Percent Purity ≥78% (Pb)
Grade Extra Pure
Packaging Glass Bottle
Chemical Name or Material Lead(II) carbonate, basic, -325 Mesh
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for b
EINECSNumber 215-290-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.