Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Plastic bottle
7758-95-4
Cl2Pb
278.11
MFCD00011157
HWSZZLVAJGOAAY-UHFFFAOYSA-L
lead dichloride,lead ii chloride,lead chloride,plumbous chloride,lead 2+ chloride,lead chloride pbcl2,pbcl2,ccris 7565,lead ii chloride, ultra dry
24459
dichlorolead
Cl[Pb]Cl
7758-95-4
Cl2Pb
278.11
MFCD00011157
HWSZZLVAJGOAAY-UHFFFAOYSA-L
lead dichloride,lead ii chloride,lead chloride,plumbous chloride,lead 2+ chloride,lead chloride pbcl2,pbcl2,ccris 7565,lead ii chloride, ultra dry
24459
dichlorolead
Cl[Pb]Cl
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula PbCl2
Merck Index 15, 5459
Solubility Information Solubility in water: 0.99g/L (20°C). Other solubilities: readly soluble in solution of nh4cl and nh4no3,soluble in alkalis
Formula Weight 278.11
Physical Form Crystalline Powder
Percent Purity 99%
Packaging Plastic bottle
Solubility Solubility in water: 0.99g/L (20°C). Other solubilities: readly soluble in solution of nh4cl and nh4no3, soluble in alkalis
Boiling Point 950.0°C
Color White
Melting Point 501.0°C
Quantity 50g
Chemical Name or Material Lead(II) chloride
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child.
Su

GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C

GHS Signal Word: Danger

EINECSNumber : 231-845-5