Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) chloride, ultra dry, 99.999% (metals basis)

Catalog No. AA3570414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3570414 25 g
AA3570406 5 g
AA3570422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3570414 Supplier Thermo Scientific Chemicals Supplier No. 03570414
Add to Cart

Used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. | CAS: 7758-95-4 | Cl2Pb | 278.10 g/mol

Lead(II) chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-95-4
Molecular Formula Cl2Pb
Molecular Weight (g/mol) 278.10
MDL Number MFCD00011157
InChI Key HWSZZLVAJGOAAY-UHFFFAOYSA-L
Synonym lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
PubChem CID 24459
IUPAC Name dichlorolead
SMILES [Cl-].[Cl-].[Pb++]

Specifications

Boiling Point 951°C
Melting Point 501°C
Physical Form Beads
Quantity 25 g
UN Number UN2291
Solubility Information Soluble in hot water,alkali hydroxides and ammonium chloride solution. Insoluble in cold water and alcohol.
Formula Weight 278.1
Percent Purity 99.999%
Packaging Ampouled under argon
Specific Gravity 5.85
Density 5.85
Chemical Name or Material Lead(II) chloride
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 231-845-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.