Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) iodide, 99%

Catalog No. AC198862500
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle

CAS: 10101-63-0 | I2Pb | 461.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-63-0
Molecular Formula I2Pb
Molecular Weight (g/mol) 461.00
MDL Number MFCD00011163
InChI Key RQQRAHKHDFPBMC-UHFFFAOYSA-L
Synonym lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane
PubChem CID 24931
IUPAC Name λ²-lead(2+) diiodide
SMILES [I-].[I-].[Pb++]

Specifications

Melting Point 402°C
Color Yellow to Orange
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula PbI2
Merck Index 15, 5466
Formula Weight 461
Percent Purity 99%
Loss on Drying 0.1% max.
Packaging Glass bottle
Specific Gravity 6.16
Density 6.1600g/mL
Chemical Name or Material Lead(II) iodide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for b
EINECSNumber 233-256-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.