Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II,IV) oxide, 97% (metals basis)

Catalog No. AA14232A7
Click to view available options
Quantity:
1 kg
5 kg

CAS: 1314-41-6 | O4Pb3 | 685.596 g/mol

Lead(II,IV) oxide is used to prepare colorless glass, faience glaze, porcelain painting flux, iron and steel coatings, rubber pigment and in glass cement. It is also used in gas and steam pipes, storage batteries, writing on glass and to make lead peroxide and matches. It is associated with linseed oil and used as a thick, long-lasting anti-corrosive paint. It gives better water resistant properties by replacing magnesium oxide. Further, it is used in the manufacture of lead glass and rustproof primer paints. In addition to this, it acts as a pigment for primer paints for iron objects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.596
MDL Number MFCD00078491
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
Synonym lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
PubChem CID 16685188
IUPAC Name 1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
SMILES O1[Pb]O[Pb]12O[Pb]O2

Specifications

Melting Point ∼500°C (decomposition)
Color Orange to Red
Physical Form Powder
Quantity 5 kg
Linear Formula Pb3O4
UN Number UN1479
Merck Index 14,5425
Solubility Information Soluble in hydrochloric acid,glacial acetic acid and nitric acid and hydrogen peroxide. Insoluble in water and alcohol.
Formula Weight 685.6
Percent Purity 97%
Odor Odorless
Density 9.1 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II,IV) oxide
Show More Show Less
Hazard Category H302+H332-H350-H360-H362-H372
Hazard Statement GHS H Statement
H360-H373-H272-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May intensify fire
oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P263-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-235-6
RTECSNumber OG5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.