Learn More
Lead(II) nitrate, 99%
Description
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.
Solubility
Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.
Notes
Incompatible with strong reducing agents, organic materials and powdered metals.
Specifications
Specifications
| Melting Point | 470°C |
| Quantity | 2500 g |
| UN Number | UN1469 |
| Merck Index | 14,5414 |
| Solubility Information | Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol. |
| Formula Weight | 331.2 |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 4.53 |
| Chemical Name or Material | Lead(II) nitrate |
Safety and Handling
| Hazard Category | H272-H302+H332-H318-H336-H350-H360Df-H373 |
| Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LEAD NITRATE |
| EINECSNumber | 233-245-9 |
| RTECSNumber | OG2100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.