Learn More
Lead(II) oxide, 99.99% (metals basis)
Description
Lead(II) oxide is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. It is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 886°C |
Color | Yellow |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in concentrated alkali,hydrochloric acid,and ammonium chloride. Insoluble in water,dilute alkali and alcohol. |
IUPAC Name | plumbanone |
Formula Weight | 223.19 |
Odor | Odorless |
Appearance | Yellow powder |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H302+H332-H350-H360Df-H372 |
missing translation for 'healthHazard3' | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | DOT Class: 9, Packaging Group: III |
EINECSNumber | 215-267-0 |
RTECSNumber | OG1750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.