Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lead(II) oxide, Puratronic™, 99.9995% (metals basis)

Catalog No. AA1072806
Click to view available options
:
5 g

Specifications

Melting Point 886°
CAS 1317-36-8
Physical Form Solid
Quantity 5 g
Molecular Formula OPb
Linear Formula PbO
MDL Number MFCD00011164
UN Number UN3077
Merck Index 14,5413
Formula Weight 223.19
Percent Purity 99.9995%
Density 9.53
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.