Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) perchlorate trihydrate, ACS, 97.0-102.0%
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100°C (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 500 g |
Linear Formula | Pb(ClO4)2·3H2O |
UN Number | UN1470 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 460.15 (406.10 Anhydrous) |
Percent Purity | ≤97% |
Grade | ACS |
Show More |
Safety and Handling
Hazard Category | H272-H302+H332-H336-H350-H360Df-H373 |
Hazard Statement | GHS H Statement H271-H360-H373-H302-H332 May cause fire or explosion strong oxidizer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LEAD PERCHLORATE, SOLID |
EINECSNumber | 237-125-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction