Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead(II) perchlorate trihydrate, ACS, 97% min

Titrant in complexometric titrations

Manufacturer:  Alfa Aesar™ 1164022

Quantity 100g

 View more versions of this product

Catalog No. AA1164022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13453-62-8
460.15 (406.10 Anhydrous)
UN1470
ACS Grade
Pb(ClO4)2·3H2O
MFCD00149817
Lead(II) perchlorate trihydrate

Specifications

13453-62-8
Pb(ClO4)2·3H2O
MFCD00149817
Soluble in water
Odorless
White
100g
≤97%
Crystalline
460.15 (406.10 Anhydrous)
UN1470
2.60g/mL
Hygroscopic
100°C (dec.)
Lead(II) perchlorate trihydrate
ACS Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H271-H360-H373-H302-H332
May cause fire or explosion
strong oxidizer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P221-P283-P210-P306+P360-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LEAD PERCHLORATE, SOLID

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit