Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) sulfate, 99%

Catalog No. p-3523473
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 7446-14-2 | O4PbS | 303.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-14-2
Molecular Formula O4PbS
Molecular Weight (g/mol) 303.25
MDL Number MFCD00011166
InChI Key KEQXNNJHMWSZHK-UHFFFAOYSA-L
Synonym lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
PubChem CID 24008
IUPAC Name lead(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 1170.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Merck Index 15, 5475
Solubility Information Solubility in water: slightly soluble
Formula Weight 303.25
Percent Purity 99%
Identification Confirmed
Packaging Plastic bottle
Chemical Name or Material Lead(II) sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a pos
EINECSNumber 231-198-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.