Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
Lead(II) sulfate is used in pigments, galvanic batteries, lithography, weighting fabrics. It is also used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1170°C |
Color | White |
Physical Form | Lump/Powder |
Quantity | 500 g |
Solubility Information | Partially soluble in water,dilute HCl and HNO3. Soluble in sodium hydroxide,ammonium acetate or tartrate,concentrated hydriodic acid. Insoluble in alcohol |
Formula Weight | 303.25 |
Grade | Reagent |
Odor | Odorless |
Appearance | White powder/lump |
Density | 6.2 |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H350-H360-H373 |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
DOTInformation | DOT Class: 9; Packaging Group: III |
EINECSNumber | 231-198-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction