Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) sulfide, Pb 82% min.
Click to view available options
Quantity:
500 g
2500 g
Description
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.
Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.
Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.
Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.
Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 1114°C |
| Quantity | 500 g |
| UN Number | UN3077 |
| Merck Index | 14,5421 |
| Solubility Information | Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide. |
| IUPAC Name | λ²-lead(2+) sulfanediide |
| Formula Weight | 239.25 |
| Concentration or Composition (by Analyte or Components) | Pb 82% min. |
| Odor | Odorless |
| Refractive Index | 3.921 |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H350-H360Df-H373 |
| Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 215-246-6 |
| RTECSNumber | OG4550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction