Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) sulfide, Pb 82% min.

Catalog No. AAA102680E
Click to view available options
Quantity:
500 g
2500 g

CAS: 1314-87-0 | PbS | 239.30 g/mol

Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(II) sulfide is used as a semiconductor. Used in electronic devices and Infrared sensor.

Solubility
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol, and potassium hydroxide.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
SMILES [S--].[Pb++]

Specifications

Melting Point 1114°C
Quantity 2500 g
UN Number UN3077
Merck Index 14,5421
Solubility Information Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide.
IUPAC Name sulfanylidenelead
Formula Weight 239.25
Concentration or Composition (by Analyte or Components) Pb 82% min.
Odor Odorless
Refractive Index 3.921
Density 7.5
Chemical Name or Material Lead (II) sulfide
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-246-6
RTECSNumber OG4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.