Learn More
Lead(II) telluride, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Lead(II) telluride is used to make thermoelectric converter (cooler, generator, heat), infrared detectors and tunnel diode. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 917°C |
Physical Form | Pieces |
Quantity | 25 g |
UN Number | UN3284 |
Merck Index | 14,5422 |
Solubility Information | Insoluble in water. |
Formula Weight | 334.79 |
Percent Purity | 99.999% |
Odor | Product specific |
Density | 8.164 g/mL |
Show More |
Safety and Handling
Hazard Category | H302+H332-H350-H360Df-H373 |
Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 215-247-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.