Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) titanium(IV) isopropoxide, 99.9% (metals basis), 5% w/v in isopropanol

Catalog No. AA39153AC
Click to view available options
Quantity:
25 mL
100 mL

Catalog No. AA39153AC

Supplier: Thermo Scientific Chemicals 039153AC

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00210621

Specifications

Color Yellow to Brown
Flash Point 12°C (54°F)
Odor Alcohol-like
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 609.29
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Lead(II) titanium(IV) isopropoxide
Show More Show Less
Hazard Category H225-H319-H336-H360Df
Hazard Statement GHS H Statement
H225-H360-H373-H302-H332-H319-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.