Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(IV) acetate, 96% (dry wt.), stab. with 5-10% glacial acetic acid

Catalog No. p-7022667
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g

Catalog No. AAA1555122

Supplier: Thermo Scientific Chemicals A1555122

Only null left
Add to Cart

CAS: 546-67-8 | C8H16O8Pb | 447.408 g/mol

Lead(IV) acetate is an important oxidizing agent and a source of acetyloxy group used in organic synthesis. For example, 1,4-dioxene is prepared from dioxane involving 2-acetoxy-1,4-dioxane as an intermediate. Similarly, it is used for the preparation of bis(trifluoromethyl)diazomethane from hexafluoroacetone hydrazone. It also reacts with alkenes, alcohols having a delta-proton and di-n-butyl d-tartrate to get gamma-lactones, cyclic ethers and n-butyl glyoxylate respectively. It induces the cleavage of 1,2-diols to the corresponding aldehydes or ketones. It is actively involved in the Kochi reaction for the decarboxylation of carboxylic acids to alkyl halides and used as an alternative reagent to bromine in the Hofmann rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lead(IV) acetate is an important oxidizing agent and a source of acetyloxy group used in organic synthesis. For example, 1,4-dioxene is prepared from dioxane involving 2-acetoxy-1,4-dioxane as an intermediate. Similarly, it is used for the preparation of bis(trifluoromethyl)diazomethane from hexafluoroacetone hydrazone. It also reacts with alkenes, alcohols having a delta-proton and di-n-butyl d-tartrate to get gamma-lactones, cyclic ethers and n-butyl glyoxylate respectively. It induces the cleavage of 1,2-diols to the corresponding aldehydes or ketones. It is actively involved in the Kochi reaction for the decarboxylation of carboxylic acids to alkyl halides and used as an alternative reagent to bromine in the Hofmann rearrangement.

Solubility
Soluble in water, ethanol, chloroform, benzene, nitrobenzene, tetrachloroethane, nitric acid, hot acetic acid and hydrochloric acid.

Notes
Air and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, strong acids and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-67-8
Molecular Formula C8H16O8Pb
Molecular Weight (g/mol) 447.408
MDL Number MFCD00008693
InChI Key NVTAREBLATURGT-UHFFFAOYSA-N
Synonym lead iv acetate
PubChem CID 50931538
IUPAC Name acetic acid;lead
SMILES CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]

Specifications

Melting Point 175°C
Density 2.28
Linear Formula Pb(OOCCH3)4
Quantity 100 g
UN Number UN1616
Beilstein 3595640
Sensitivity Moisture sensitive
Merck Index 14,5423
Solubility Information Soluble in water,ethanol,chloroform,benzene,nitrobenzene,tetrachloroethane,nitric acid,hot acetic acid and hydrochloric acid.
Formula Weight 443.37
Percent Purity 96%
Assay (dry wt.)
Chemical Name or Material Lead(IV) acetate, stabilized with 5 to 10% glacial acetic acid
Show More Show Less
Hazard Category H302+H332-H314-H335-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332-H201
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Explosive; mass explosion hazard.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber 208-908-0
RTECSNumber AI5300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.