Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(IV) acetate, 95%, stabilized, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
546-67-8,64-19-7
C8H12O8Pb
443.36
NVTAREBLATURGT-UHFFFAOYSA-N
lead iv acetate
50931538
acetic acid;lead
CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]
546-67-8,64-19-7
C8H12O8Pb
443.36
NVTAREBLATURGT-UHFFFAOYSA-N
lead iv acetate
50931538
acetic acid;lead
CC(=O)O.CC(=O)O.CC(=O)O.CC(=O)O.[Pb]
Oxidation reagent
Assay Percent Range 94% min. (Iodometry)
Linear Formula (CH3CO2)4Pb
Fieser 01,537; 02,234; 03,168; 04,278; 05,365; 06,313; 08,269; 09,265; 10,228; 12,270; 13,62; 14,188; 16,193
Merck Index 15,5478
Solubility Information Solubility in water: decomposes in water forming pb02 and acetic acid. Other solubilities: soluble in hot glacial acetic acid,soluble in benzene,chloroform,tetrachloroethane,,toluene,acetonitrile,nitrobenzene and,dichloromethane
Formula Weight 443.36
Physical Form Crystalline Powder
Percent Purity 95%
Packaging Glass bottle
Solubility Solubility in water: decomposes in water forming pb02 and acetic acid. Other solubilities: soluble in hot glacial acetic acid, soluble in benzene, chloroform, tetrachloroethane,, toluene, acetonitrile, nitrobenzene and, dichloromethane
Stabilizer 4 to 5% (ca.) acetic acid
Color Orange-Pink to White
Melting Point 175.0°C to 180.0°C
Quantity 100g
Chemical Name or Material Lead(IV) acetate
Show More Show Less

GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Harmful if inhaled.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 208-908-