Learn More
Invitrogen™ Lead Monoclonal Antibody (PB1)

Description
MA1-10880 detects Lead. MA1-10880 has been successfully used in ELISA applications. The MA1-10880 immunogen is Lead, conjugated to KLH.
Specifications
Specifications
| Antigen | Lead |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | PB1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with no preservative |
| Gene Alias | Lead metal; Pb; Plumbum |
| Host Species | Mouse |
| Immunogen | Lead, conjugated to KLH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.