Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate Stock Solution, 100 ppm Pb, Reagents

Catalog No. p-200022072
Click to view available options
Quantity:
500 mL
1 L

Lead Nitrate Stock Solution, 100 ppm Pb, Reagent, use for USP lead testing. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Lead Nitrate Stock Solution, 100 ppm Pb
Packaging Glass Bottle
CAS 7732-18-5, 7697-37-2
Specific Gravity 1.01
Physical Form Liquid
Boiling Point 100°C
Synonym Plumbous Nitrate, Lead(II) Nitrate, Lead Dinitrate
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.