Learn More
Meridian Bioscience™ LeadCare™ II Analyzer
Shop All Meridian Bioscience Products

Description
Includes
Portable blood-lead testing instrument, AC power cord, AA batteries, User’s Guide (English), Quick Reference Guide, Instructional video, and CDC blood-collection video
- Safe, fast and inexpensive to use
- Simple to operate
- Small, lightweight and portable
- Accurate reporting
- One-touch electronic calibration
- Reagents stable at room temperature
- FDA cleared
Specifications
- Power Source: 4 AA batteries or AC adapter
- Dimensions: 22.8 x 16.5 x 8.9cm (9 x 6.5 x 3.5 in.)
- Weight: 1 kg (2.4 lb.)
Certifications
CE, UL, CSA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Type | Analyzer |
Sample Type | Capillary Whole Blood |
For Use With (Application) | Quantitative blood lead results in just three minutes |
Calibration | One-touch electronic calibration |
Certifications/Compliance | UL, CSA |
Includes | Portable blood-lead testing instrument, AC power cord, AA batteries, User’s Guide (English), Quick Reference Guide, Instructional video, and CDC blood-collection video |
Electrical Requirements | 4 AA batteries or AC adapter |
Weight (English) | 2.4 lb. |
Weight (Metric) | 1 kg |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.