Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LECT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107102 Supplier Thermo Scientific Supplier No. PA5107102
Add to Cart
Add to Cart

LECT1 Polyclonal Antibody

LECT1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LECT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene CNMD
Gene Accession No. O75829
Gene Alias BRICD3; BRICHOS domain containing 3; CHM1; Chm-1; Chmi; CHM-I; Chondromodulin; Chondromodulin 1; chondromodulin I; chondromodulin-1; Chondromodulin-I; Chondrosurfactant protein; CH-SP; Cnmd; LECT1; leukocyte cell derived chemotaxin 1; leukocyte cell-derived chemotaxin 1; multiple myeloma tumor suppressor 1; MYETS1
Gene Symbols CNMD
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human LECT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.