Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LECT2 Antibody (OTI2A11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271122AF700

Catalog No. N271122A700


Only null left
Add to Cart

Description

Description

LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

LECT2
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
LECT2
Recombinant protein fragment corresponding to amino acids 19-151 of human LECT2 (NP_002293) produced in E.coli.
0.1 mL
3950.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI2A11
Western Blot, Immunohistochemistry
chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628
Mouse
Affinity Purified
Primary
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.