Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LECT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570639
Description
This target displays homology in the following species: Guinea Pig: 79%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
This gene encodes a secreted, 16 kDa protein that acts as a chemotactic factor to neutrophils and stimulates the growth of chondrocytes and osteoblasts. This protein has high sequence similarity to the chondromodulin repeat regions of the chicken myb-induced myeloid 1 protein. A polymorphism in this gene may be associated with rheumatoid arthritis.
Specifications
LECT2 | |
Polyclonal | |
Unconjugated | |
LECT2 | |
chm2; ChM-II; chondromodulin II; chondromodulin-II; hLECT2; Lect2; LECT-2; leukocyte cell derived chemotaxin 2; leukocyte cell-derived chemotaxin 2; leukocyte cell-derived chemotaxin-2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3950 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14960 | |
LECT2 | |
synthetic peptide directed towards the middle region of human LECT2 (aa 6-55). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction