Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lectin LCA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987264
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7264 Supplier Bioss Supplier No. BS6439R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lens culinaris agglutinin is composed of four subunits - two of about 17 kDa and two of 8 kDa. LCA recognizes sequences containing _-linked mannose residues but recognizes additional sugars as part of the receptor structure, giving it a narrower specificity than Con A. An _-linked fucose residue attached to the N-acetylchitobiose portion of the core oligosaccharide markedly enhances affinity. LCA has been found to be one of the most effective agents in preventing skin allograft rejection in model systems. LCA has been used to purify numerous glycoproteins, including immunoglobulins, histocompatibility antigens, and _2-macroglobulin.

Specifications

Antigen Lectin LCA
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTPRC
Gene Accession No. P08575
Gene Symbols PTPRC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Lens culinaris Lectin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.