Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEF1 (Alternate Exon) Monoclonal Antibody (1C3)
GREENER_CHOICE

Catalog No. 01674335
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-335 Supplier Invitrogen™ Supplier No. MA112487
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LEF1 (Alternate Exon) Monoclonal Antibody for Western Blot

Lymphoid enhancer-binding factor-1 (LEF1) is a 48-kD nuclear protein that is expressed in pre-B and T cells. It binds to a functionally important site in the T-cell receptor-alpha (TCRA; MIM 186880) enhancer and confers maximal enhancer activity. LEF1 belongs to a family of regulatory proteins that share homology with high mobility group protein-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF1 (Alternate Exon)
Applications Western Blot
Classification Monoclonal
Clone 1C3
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene LEF1
Gene Accession No. Q9UJU2
Gene Alias DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; LEF-1; lymphoid enhancer binding factor 1; lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3
Gene Symbols LEF1
Host Species Mouse
Immunogen Recombinant full-length LEF-1 protein.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.