Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEF1 Antibody (1C3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP266625
Description
LEF1 Monoclonal specifically detects LEF1 in Human samples. It is validated for Western Blot.Specifications
LEF1 | |
Monoclonal | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA | |
Mouse | |
Protein A or G purified | |
DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway | |
51176 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
1C3 | |
Western Blot | |
Q9UJU2 | |
LEF1 | |
Recombinant full-length LEF1 protein | |
100 μg | |
Primary | |
Human, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction