Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEF1 Rabbit anti-Human, Clone: LEF1/341R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
Antigen | LEF1 |
---|---|
Clone | LEF1/341R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
LEF1 Monoclonal specifically detects LEF1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
LEF1 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway | |
10 mM PBS with 0.05% BSA | |
51176 | |
IgG κ | |
Protein A or G purified |
LEF1/341R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA | |
Synthetic peptide corresponding to residues within aa100-200 of LEF1 | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title