Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF1 Rabbit anti-Human, Clone: SP308, Abnova™

Catalog No. 89035367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-367 Supplier Abnova Corporation Supplier No. MAB15153

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against synthetic peptide of human LEF1.

  • High specificity
  • High reactivity

Specifications

Antigen LEF1
Applications Flow Cytometry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP308
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human LEF1.
Dilution Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene Alias DKFZp586H0919/TCF1ALPHA
Gene Symbols LEF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human LEF1.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.