Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Left Access Port (50 to 125 mm)
SDP

Catalog No. 08111064 Shop All BINDER Inc. Products
Click to view available options
Diameter (Metric) Inside:
50 mm

For Use With: KBF 240, KBF-S 240, KBF-720, KBF-S 720, KBF 1020, KBF 115

  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers

Specifications

Description Left position
For Use With (Equipment) KBF 240, KBF-S 240, KBF-720, KBF-S 720, KBF 1020, KBF 115
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 50 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.