Learn More
MSA™ Left/Right™ Ear Muffs
Ear Muffs have dedicated right and left ear cups that are designed to fit ears of all shapes and sizes which offers increased comfort and protection. The headband's breakthrough design simulates the head's curves to provide a comfortable and stable fit, balancing muffs with ear cup angle.
$69.25 - $77.00
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class C
Cushion Replacements Yes
High Visibility No
Corded No
View More Specs
Products 3
Catalog Number Mfr. No. Noise Reduction Rating (NRR) Price Quantity & Availability  
Catalog Number Mfr. No. Noise Reduction Rating (NRR) Price Quantity & Availability  
19-030-081
View Documents
MSA™
10251221
33 dB
Each for $77.00
Only null left
Add to Cart
19-030-082
View Documents
MSA™
10251224
28 dB
Each for $73.00
Only null left
Add to Cart
19-030-083
View Documents
MSA™
10251226
24 dB
Each for $69.25
Only null left
Add to Cart
Description
Description
  • Precise fit provides maximum loud noise protection using minimal cup size.
  • Headband adjusts up and down to provide custom fit for different head sizes.
  • Available in low, medium, and high attenuation
Specifications
Specifications
Yes
Yes
No
Reusable
Black
Over-the-head
EN 352-1
Class C
No
Headband Ear Muff
No
Yes
No
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.