Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ left/RIGHT™ Cap-Mounted Ear Muff

Catalog No. 10129085 Shop All MSA Products
Change view
Click to view available options
Cap-Mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
10-129-085 Cap-Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-129-085 Supplier MSA™ Supplier No. 10087429

left/RIGHT™ Cap-Slotted Ear Muffs feature cups designed to account for the asymmetrical position of each ear on the head.

  • Dedicated right and left ear cups designed to fit ears of all shapes and sizes
  • For use with MSA slotted caps
  • Cups can be adjusted to the angle of the ear and independently raised or lowered
  • Blue

Specifications

Color Blue
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Equipment) MSA™ Slotted Hard Hats
For Use With (Application) Utilities, construction, general industry
High Visibility No
Model 10087429
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type left/RIGHT
Dielectric No
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.