Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEFTY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519507
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519507 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519507 Supplier Invitrogen™ Supplier No. PA519507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-19507 targets Lefty in IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-19507 immunogen is synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human Lefty. PA5-19507 detects Lefty which has a predicted molecular weight of approximately 44 kDa.

This gene encodes a member of the TGF-beta family of proteins. A similar secreted protein in mouse plays a role in left-right asymmetry determination of organ systems during development. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEFTY
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene LEFTY1
Gene Accession No. O75610, Q64280
Gene Alias AI450052; Ebaf; endometrial bleeding-associated factor; left right determination factor 1; LEFTA; Leftb; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor B; lefty; Lefty protein; LEFTY1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; TGFB4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317
Gene Symbols LEFTY1
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human Lefty.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10637, 13590
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.