Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEFTY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590431 Supplier Invitrogen™ Supplier No. PA590431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RFSQSFREVA GRFLALEAST HLLVFGMEQR LPPNSELVQA VLRLFQEPVP KAALHRHGRL SPRSARARVT VEWLRVRDDG SNRTSLIDSR LVSVHESGWK AFDVTEAVNF WQQLSRPRQP LLLQVSVQRE HLGPLASGAH KLVRFASQGA PAGLGEPQLE LHTLDLGDYG AQGDCDPEAP MTEGTRCCRQ EMYIDLQGMK WAENWVLEPP GFLAYECVGT CRQPPEALAF KWPFLGPRQC IASETDSLPM IVSIKEGGRT RPQVVSLPNM RVQKCSCASD GALVPRRLQP; Positive Samples: HT-29, SW480, BxPC-3, Mouse pancreas, Mouse spleen, Rat pancreas.

This gene encodes a member of the TGF-beta family of proteins. A similar secreted protein in mouse plays a role in left-right asymmetry determination of organ systems during development. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEFTY
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LEFTY1
Gene Accession No. O75610, Q64280
Gene Alias AI450052; Ebaf; endometrial bleeding-associated factor; left right determination factor 1; LEFTA; Leftb; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor B; lefty; Lefty protein; LEFTY1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; TGFB4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317
Gene Symbols LEFTY1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 77-366 of human LEFTY1 (NP_0662771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10637, 13590
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.