Learn More
Invitrogen™ LEFTY Polyclonal Antibody

Description
Immunogen sequence: AQGDCDPEAP MTEGTRCCRQ EMYIDLQGMK WAENWVLEPP GFLAYECVGT C Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 88%.
Specifications
Specifications
| Antigen | LEFTY |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | LEFTY1 |
| Gene Accession No. | O75610 |
| Gene Alias | AI450052; Ebaf; endometrial bleeding-associated factor; left right determination factor 1; LEFTA; Leftb; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor B; lefty; Lefty protein; LEFTY1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; TGFB4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317 |
| Gene Symbols | LEFTY1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.