Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lefty Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981194
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1194 Supplier Bioss Supplier No. BS11236R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the TGF-beta family of proteins. A similar secreted protein in mouse plays a role in left-right asymmetry determination of organ systems during development. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene.

Specifications

Antigen Lefty
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LEFTY1
Gene Accession No. O75610
Gene Alias AI450052; Ebaf; left right determination factor 1; LEFTB; Left-right determination factor 1; Left-right determination factor B; left-right determination, factor B; Lefty; Lefty protein; LEFTY1; lefty-1; LEFTYB; Protein lefty-1; Protein lefty-B; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; Tgfb4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317
Gene Symbols LEFTY1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Lefty.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10637
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.