Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lefty1/2 Rabbit anti-Human, Mouse, Rat, Clone: 2Y10P2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31611820UL
Description
Lefty1/2 Monoclonal antibody specifically detects Lefty1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Lefty1/2 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
7044 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2Y10P2 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Beta-4 (Endometrial Bleeding-Associated Factor; LEFTY A), EBAF, endometrial bleeding associated factor (left-right determination, Endometrial Bleeding Associated Factor (Left-Right Determination, Factor A;, Endometrial Bleeding-Associated Factor, LEFTA, Left-Right Determination Factor 2, Left-Right Determination Factor A, LEFTY2, LEFTYA, Protein Lefty-2, Protein Lefty-A, TGFB4, TGF-beta-4, Transforming Growth Factor, Transforming Growth Factor Beta-4 | |
A synthetic peptide corresponding to a sequence within amino acids 267-366 of human Lefty1/2 (O75610). | |
20 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction