Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEFTY2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521367

Catalog No. PIPA521367


Only null left
Add to Cart

Description

Description

Recommended positive controls: A431. Predicted reactivity: Mouse (84%), Rat (83%), Pig (83%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LEFTY2
Polyclonal
Unconjugated
LEFTY2
6030463A22Rik; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; LEFTA; Leftb; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; LEFTY2; LEFTYA; MGC46222; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; TGFB4; TGF-beta-4; Transforming growth factor beta-4
Rabbit
Antigen affinity chromatography
RUO
7044
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.01 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O00292
LEFTY2
Recombinant fragment corresponding to a region within amino acids 25 and 366 of LEFTY2 (Uniprot ID#O00292).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.